Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277742.1 | NP_001264671.1 | 5 | 3372 | ||
NM_019885.3 | NP_063938.1 | 6 | 3597 | |||
XM_005264433.4 | XP_005264490.1 | 6 | 3561 | |||
XM_011532988.1 | XP_011531290.1 | 5 | 3158 | |||
GenBank mRNA | AF252297.1 | - | 6 | 3569 |