Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 10 | 1129 | ||
NM_007700.2 | NP_031726.2 | 10 | 1129 | |||
GenBank mRNA | AK018671.1 | - | 10 | 1108 | ||
AK035326.1 | - | 10 | 1086 | |||
AK150039.1 | - | 10 | 1108 | |||
AK155328.1 | - | 10 | 1109 | |||
AK155393.1 | - | 10 | 1130 | |||
AK164236.1 | - | 10 | 1130 | |||
BC018243.1 | - | 10 | 1121 | 4511275 | ||
U12473.1 | - | 10 | 1097 |