Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168655.1 | NP_001162126.1 | 3 | 645 | ||
NM_001168656.1 | NP_001162127.1 | 3 | 603 | |||
NM_001168657.1 | NP_001162128.1 | 2 | 524 | |||
NM_007706.4 | NP_031732.1 | 3 | 672 | |||
GenBank mRNA | AK033206.1 | - | 1 | 72 | ||
AK137403.1 | - | 3 | 552 | |||
AK144810.1 | - | 3 | 614 | |||
AK160537.1 | - | 3 | 629 | |||
AK162591.1 | - | 3 | 637 | |||
BC106153.1 | - | 3 | 643 | 4989433 | ||
U88327.1 | - | 2 | 432 |