Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007737.2 | NP_031763.2 | 2 | 607 | ||
GenBank mRNA | AK084285.1 | - | 2 | 389 | ||
AK084803.1 | - | 2 | 244 | |||
AK132413.1 | - | 2 | 389 | |||
AK139130.1 | - | 2 | 182 | |||
AK147220.1 | - | 2 | 389 | |||
AK147328.1 | - | 2 | 608 | |||
AK151929.1 | - | 2 | 391 | |||
AK160008.1 | - | 2 | 181 | |||
BC055077.1 | - | 2 | 358 | 30059991 | ||
L02918.1 | - | 2 | 239 |