Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007739.2 | NP_031765.2 | 3 | 501 | ||
GenBank mRNA | AK018742.1 | - | 3 | 480 | ||
AK032048.1 | - | 3 | 480 | |||
AK046370.1 | - | 3 | 480 | |||
AK159542.1 | - | 3 | 476 | |||
BC011061.1 | - | 3 | 468 | 3674027 | ||
BC058281.1 | - | 3 | 484 |