Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007741.2 | NP_031767.2 | 11, 12 | 722 | ||
GenBank mRNA | BC029697.1 | - | 11, 12 | 661 | 5355825 | |
BU512853.1 | - | 11, 12 | 623 | |||
BY088510.1 | - | 8, 9 | 357 |