Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111062.1 | NP_001104532.1 | 3, 4 | 694 | ||
NM_001111063.1 | NP_001104533.1 | 2, 3 | 505 | |||
NM_007744.3 | NP_031770.2 | 2, 3 | 637 | |||
GenBank mRNA | AF076156.1 | - | 2, 3 | 439 | ||
AK148311.1 | - | 3, 4 | 361 | |||
BC010402.1 | - | 2, 3 | 431 | 4210097 | ||
CB946852.1 | - | 2, 3 | 474 | |||
CX237493.1 | - | 2, 3 | 637 |