Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007760.3 | NP_031786.2 | 2 | 687 | ||
GenBank mRNA | AK041431.1 | - | 2 | 462 | ||
AK132179.1 | - | 3 | 514 | |||
AK170740.1 | - | 2 | 473 | |||
AK218779.1 | - | 2 | 295 | |||
BC006668.1 | - | 2 | 434 | 3499169 | ||
X85983.1 | - | 2 | 402 |