Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm37013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174154.2 | NP_001167625.1 | 1 | 563 |
Gene Symbol: Pcdha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007766.2 | NP_031792.1 | 1 | 522 | ||
GenBank mRNA | AK048980.1 | - | 1 | 553 | ||
AK149282.1 | - | 1 | 564 | |||
AY013762.1 | - | 1 | 445 | |||
BC060211.1 | - | 1 | 536 | 6838078 | ||
D86916.1 | - | 1 | 522 |