Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301282.1 | NP_001288211.1 | 3 | 827 | ||
NM_001301283.1 | NP_001288212.1 | 3 | 824 | |||
NM_007795.1 | NP_031821.1 | 3 | 786 | |||
GenBank mRNA | BC024381.1 | - | 3 | 778 | 4986446 | |
U18366.1 | - | 3 | 786 |