Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290825.1 | NP_001277754.1 | 22 | 2085 | ||
NM_007732.2 | NP_031758.2 | 22 | 2085 | |||
GenBank mRNA | BC125031.1 | - | 22 | 2085 | 40086689 | |
BC125032.1 | - | 22 | 2084 | 40086691 | ||
L08407.1 | - | 22 | 2137 |