Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007738.3 | NP_031764.2 | 105, 106 | 7856 | ||
GenBank mRNA | CX224636.1 | - | 8, 9 | 456 | ||
U32107.1 | - | 105, 106 | 7856 |