Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007741.2 | NP_031767.2 | 24 | 1424 | ||
GenBank mRNA | AA171281.1 | - | 4 | 238 | ||
BC029697.1 | - | 24 | 1363 | 5355825 | ||
CJ046324.1 | - | 8 | 399 |