Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007762.4 | NP_031788.1 | 11 | 1220 | ||
GenBank mRNA | AB375515.1 | - | 11 | 1030 | ||
AF483484.1 | - | 11 | 1030 | |||
AF483485.1 | - | 11 | 1030 | |||
AK144430.1 | - | 11 | 1288 | |||
AK158806.1 | - | 11 | 1221 | |||
BC103675.1 | - | 11 | 1048 | 40046992 | ||
BC104735.1 | - | 12 | 1048 | 40046995 | ||
BC105673.1 | - | 12 | 1124 | 40046994 | ||
BC105675.1 | - | 11 | 1048 | 40046993 | ||
FJ668671.1 | - | 10 | 910 | |||
X72305.1 | - | 11 | 1061 |
Set Membership: |
G-protein coupled receptor |