Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037859.2 | NP_001032948.2 | 12 | 1956 | ||
GenBank mRNA | AK004947.1 | - | 11 | 1766 | ||
AK041299.1 | - | 11 | 1744 | |||
AK076215.1 | - | 11 | 1764 | |||
AK079247.1 | - | 11 | 1773 | |||
AK080326.1 | - | 11 | 1754 | |||
AK143545.1 | - | 11 | 1757 | |||
AK146069.1 | - | 12 | 1957 | |||
AK154128.1 | - | 11 | 1771 | |||
AK154556.1 | - | 11 | 1767 | |||
AK154653.1 | - | 11 | 1767 | |||
AK154699.1 | - | 11 | 1767 | |||
AK155488.1 | - | 11 | 1777 | |||
AK155565.1 | - | 11 | 1786 | |||
AK155576.1 | - | 1773 | ||||
AK155640.1 | - | 11 | 1771 | |||
AK155654.1 | - | 11 | 1771 | |||
AK155858.1 | - | 11 | 1774 | |||
AK156550.1 | - | 11 | 1791 | |||
AK171386.1 | - | 11 | 1771 | |||
BC036343.1 | - | 11 | 1743 | 4948135 | ||
BC043054.1 | - | 11 | 1747 | 6418718 | ||
BC050024.1 | - | 11 | 1854 | |||
BC066863.1 | - | 12 | 1865 | 30436119 | ||
X06368.1 | - | 11 | 1749 | |||
X68932.1 | - | 11 | 1659 |