Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144063.1 | NP_001137535.1 | 10, 11 | 1288 | ||
NM_020896.3 | NP_065947.1 | 11, 12 | 1492 | |||
NM_145638.2 | NP_663613.1 | 10, 11 | 1281 | |||
XM_011519873.2 | XP_011518175.1 | 11, 12 | 1831 | |||
XM_017017163.1 | XP_016872652.1 | 11, 12 | 1839 | |||
XM_017017164.1 | XP_016872653.1 | 6, 7 | 1410 | |||
GenBank mRNA | AB074006.1 | - | 11, 12 | 1433 | ||
AF392453.1 | - | 11, 12 | 1405 | |||
AF410430.1 | - | 10, 11 | 1350 | |||
AK055617.1 | - | 6, 7 | 826 | |||
AK056510.1 | - | 11, 12 | 1450 | |||
AK293000.1 | - | 10, 11 | 1237 | |||
AK297475.1 | - | 7, 8 | 587 | |||
AK301003.1 | - | 9, 10 | 1128 | |||
AK303205.1 | - | 11, 12 | 1310 | |||
AK309411.1 | - | 11, 12 | 1470 | |||
AL136918.1 | - | 11, 12 | 1405 | |||
BC017505.1 | - | 2, 3 | 148 | |||
BC032646.1 | - | 10, 11 | 1202 | 5518491 | ||
BC039579.1 | - | 11, 12 | 1403 | 5769002 |