Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007805.4 | NP_031831.2 | 6 | 2459 | ||
GenBank mRNA | AK014395.2 | - | 6 | 2460 | ||
AK028780.1 | - | 5 | 2333 | |||
AK158978.1 | - | 7 | 2790 | |||
AK169711.1 | - | 6 | 2448 | |||
BC006732.1 | - | 6 | 2499 | 3963612 |