Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007868.5 | NP_031894.1 | 3 | 421 | ||
GenBank mRNA | AK044536.1 | - | 3 | 221 | ||
AK052856.1 | - | 3 | 422 | |||
M68859.1 | - | 3 | 394 |