Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007881.4 | NP_031907.2 | 4 | 570 | ||
GenBank mRNA | BC050920.1 | - | 4 | 570 | 6406687 | |
BC053051.1 | - | 4 | 398 | 5706586 | ||
D87744.1 | - | 3 | 280 |