Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp17la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007887.2 | NP_031913.1 | 3 | 272 | ||
GenBank mRNA | AY148453.1 | - | 2 | 101 | ||
BC140988.1 | - | 3 | 301 | 9055600 | ||
U41636.1 | - | 3 | 284 |