Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007896.3 | NP_031922.1 | 5, 6 | 741 | ||
GenBank mRNA | AK013626.1 | - | 5, 6 | 681 | ||
AK146206.1 | - | 5, 6 | 659 | |||
AK154245.1 | - | 5, 6 | 685 | |||
BC043333.1 | - | 5, 6 | 705 | |||
BC052405.1 | - | 4, 5 | 616 | |||
BC052728.1 | - | 652 | ||||
BC064444.1 | - | 652 | 6815724 | |||
U51196.1 | - | 5, 6 | 671 |