Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128932.1 | NP_001122404.1 | 4 | 404 | ||
NM_021187.3 | NP_067010.3 | 3 | 826 | |||
GenBank mRNA | AF236085.1 | - | 3 | 476 | ||
AK125714.1 | - | 1 | 653 | |||
AK289424.1 | - | 3 | 394 | |||
AL833940.1 | - | 3 | 376 | |||
BC016853.1 | - | 4 | 404 | 3846027 | ||
BU180723.1 | - | 3 | 237 |