Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007911.5 | NP_031937.1 | 2 | 567 | ||
GenBank mRNA | AF025288.1 | - | 2 | 185 | ||
AK048305.1 | - | 2 | 511 | |||
BC052001.1 | - | 2 | 384 | 5695429 | ||
BC058617.1 | - | 2 | 500 | 5695418 |