Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110826.1 | NP_001104296.1 | 4, 5 | 743 | ||
NM_007841.4 | NP_031867.1 | 4, 5 | 686 | |||
NM_181324.3 | NP_851841.2 | 4, 5 | 711 | |||
GenBank mRNA | AF038995.1 | - | 3, 4 | 305 | ||
AK036648.1 | - | 4, 5 | 739 | |||
AK054144.1 | - | 4, 5 | 705 | |||
AK148483.1 | - | 4, 5 | 708 | |||
BC021452.1 | - | 3, 4 | 603 | 5043599 | ||
CD353843.1 | - | 4, 5 | 673 |