Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007859.4 | NP_031885.3 | 3, 4 | 539 | ||
GenBank mRNA | AB009377.1 | - | 3, 4 | 447 | ||
AK077619.1 | - | 3, 4 | 585 | |||
AK089267.1 | - | 3, 4 | 516 | |||
BC053052.1 | - | 3, 4 | 469 | 6403143 |