Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007861.5 | NP_031887.2 | 3 | 263 | ||
GenBank mRNA | AK014092.1 | - | 3 | 206 | ||
AK117104.1 | - | 3 | 207 | |||
AK135807.1 | - | 3 | 206 | |||
AK136193.1 | - | 3 | 207 | |||
AK153399.1 | - | 3 | 207 | |||
AK167885.1 | - | 3 | 207 | |||
AK168157.1 | - | 3 | 228 | |||
AK168875.1 | - | 3 | 148 | |||
BC003368.1 | - | 3 | 190 | 3589344 | ||
BY251665.1 | - | 3 | 230 | |||
BY282701.1 | - | 3 | 266 | |||
U73445.1 | - | 3 | 240 |