Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007865.3 | NP_031891.2 | 2 | 758 | ||
GenBank mRNA | BC057400.1 | - | 2 | 271 | 6402691 | |
BC065063.1 | - | 2 | 522 | 6416191 | ||
X80903.1 | - | 2 | 188 |