Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007870.3 | NP_031896.3 | 1 | 328 | ||
GenBank mRNA | AF047355.1 | - | 1 | 303 | ||
AK156109.1 | - | 161 | ||||
AK171843.1 | - | 1 | 161 | |||
AK172254.1 | - | 1 | 721 | |||
BC012671.1 | - | 1 | 281 | 4160709 | ||
U76110.1 | - | 1 | 137 |