Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243645.1 | NP_001230574.1 | 4 | 2287 | ||
NM_021914.7 | NP_068733.1 | 4 | 2310 | |||
NM_138638.4 | NP_619579.1 | 4 | 2452 | |||
NR_028130.1 | 4 | 2309 | ||||
NR_028131.1 | 4 | 2198 | ||||
XM_011536363.2 | XP_011534665.1 | 4 | 2248 | |||
GenBank mRNA | AF134802.1 | - | 4 | 2319 | ||
AF134803.1 | - | 4 | 2221 | |||
AL117457.1 | - | 1 | 1534 |