Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007831.3 | NP_031857.2 | 10, 11 | 2342 | ||
GenBank mRNA | AK157715.1 | - | 10, 11 | 2291 | ||
X85788.1 | - | 7, 8 | 1892 |
Set Membership: |
G-protein coupled receptor |