Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109752.1 | NP_001103222.1 | 9 | 1317 | ||
NM_007864.3 | NP_031890.1 | 9 | 1326 | |||
GenBank mRNA | BC014807.1 | - | 9 | 1062 | 4501403 | |
BQ959940.1 | - | 3 | 469 | |||
D50621.1 | - | 9 | 1068 |