Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007885.2 | NP_031911.1 | 4 | 3641 | ||
GenBank mRNA | D42049.1 | - | 4 | 3511 |