Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021961.5 | NP_068780.2 | 3 | 792 | ||
GenBank mRNA | AL035973.1 | - | 1 | 77 | ||
AL833289.1 | - | 2 | 326 | |||
BC115398.1 | - | 2 | 285 | 40068609 | ||
DR000961.1 | - | 3 | 630 |