Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004620.1 | - | 5 | 1005 | 3585323 | |
BC004703.1 | - | 5 | 1637 | 3499265 | ||
DV661455.1 | - | 4 | 601 | |||
X97755.1 | - | 5 | 1003 |