Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083592.1 | NP_001077061.1 | 3 | 718 | ||
NM_005012.3 | NP_005003.2 | 3 | 718 | |||
XM_011541526.1 | XP_011539828.1 | 3 | 317 | |||
XM_017001376.1 | XP_016856865.1 | 2 | 618 | |||
XM_017001377.1 | XP_016856866.1 | 5 | 3261 | |||
GenBank mRNA | AK308088.1 | - | 3 | 724 | ||
BC080541.1 | - | 3 | 718 | |||
BC128386.1 | - | 4 | 328 | |||
M97675.1 | - | 3 | 694 |