Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007963.2 | NP_031989.2 | 4 | 1085 | ||
GenBank mRNA | AJ001482.1 | - | 2 | 241 | ||
AK033517.1 | - | 4 | 519 | |||
AK051098.1 | - | 4 | 942 | |||
AK085548.1 | - | 4 | 1086 | |||
BC076620.1 | - | 4 | 1037 | |||
BC139762.1 | - | 2 | 203 | 40109903 | ||
JQ665270.1 | - | 2 | 233 |