Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007964.2 | NP_031990.2 | 2 | 142 | ||
GenBank mRNA | AK134294.1 | - | 2 | 343 | ||
AK162905.1 | - | 2 | 402 | |||
AK165332.1 | - | 3 | 135 | |||
AK165885.1 | - | 2 | 95 | |||
BC030056.1 | - | 3 | 149 | |||
BI737814.1 | - | 2 | 66 | |||
U53586.1 | - | 2 | 142 |