Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2rl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007975.3 | NP_032001.2 | 2 | 1117 | ||
GenBank mRNA | AF080215.1 | - | 2 | 1094 | ||
AK036427.1 | - | 2 | 1118 | |||
AK153720.1 | - | 2 | 1111 | |||
AK156729.1 | - | 2 | 1118 | |||
BC119461.2 | - | 1 | 1357 | 40045612 |