Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007978.3 | NP_032004.2 | 1 | 102 | ||
GenBank mRNA | AF299331.1 | - | 1 | 131 | ||
AK033184.1 | - | 1 | 104 | |||
AK046025.1 | - | 1 | 117 | |||
AK046459.1 | - | 1 | 117 | |||
AK054140.1 | - | 1 | 103 | |||
AK166334.1 | - | 1 | 118 | |||
BC131938.1 | - | 1 | 130 | 40130215 | ||
BC131940.1 | - | 1 | 130 | 40130217 | ||
M83118.1 | - | 1 | 386 |