Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007989.4 | NP_032015.1 | 1 | 217 | ||
GenBank mRNA | AB221574.1 | - | 1 | 144 | ||
AF069303.1 | - | 1 | 143 | |||
AF079514.1 | - | 1 | 140 | |||
AF110506.1 | - | 1 | 140 | |||
AF177770.1 | - | 1 | 140 | |||
BC119156.1 | - | 1 | 217 | 8733905 | ||
BC137781.1 | - | 1 | 217 | 8860800 |