Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007997.1 | NP_032023.1 | 2 | 175 | ||
GenBank mRNA | AK134068.1 | - | 2 | 166 | ||
BC057146.1 | - | 2 | 109 | |||
BC071268.1 | - | 2 | 175 | 6517167 | ||
D49920.1 | - | 2 | 175 |