Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008014.3 | NP_032040.1 | 1, 2 | 251 | ||
GenBank mRNA | AK180043.1 | - | 1, 2 | 39 | ||
AK182937.1 | - | 1, 2 | 39 | |||
AK183594.1 | - | 1, 2 | 39 | |||
AK185279.1 | - | 1, 2 | 39 | |||
BC009004.1 | - | 1, 2 | 251 | 3499081 | ||
U42383.1 | - | 1, 2 | 137 |