Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002777.1 | - | 5 | 489 | ||
AK019362.1 | - | 5 | 517 | |||
AK145400.1 | - | 5 | 506 | |||
AK145986.1 | - | 4 | 539 | |||
AK151047.1 | - | 3 | 300 | |||
AK154751.1 | - | 5 | 461 | |||
AK168333.1 | - | 5 | 497 | |||
AK169186.1 | - | 5 | 420 | |||
AK169242.1 | - | 5 | 504 | |||
BC004671.1 | - | 5 | 464 | 3498682 | ||
CX239711.1 | - | 6 | 593 | |||
X60203.1 | - | 5 | 479 |