Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007943.3 | NP_031969.1 | 3 | 230 | ||
GenBank mRNA | AY190610.1 | - | 3 | 203 | ||
BB858255.1 | - | 3 | 232 | |||
BC048783.1 | - | 3 | 204 | 6466448 | ||
L21768.1 | - | 3 | 203 |