Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122832.1 | NP_001116304.1 | 4 | 241 | ||
NM_001289859.1 | NP_001276788.1 | 4 | 241 | |||
NM_007944.3 | NP_031970.2 | 4 | 241 | |||
GenBank mRNA | AK036662.1 | - | 4 | 207 | ||
AK036728.1 | - | 4 | 219 | |||
AK079132.1 | - | 4 | 209 | |||
AK146781.1 | - | 4 | 219 | |||
AK152602.1 | - | 4 | 219 | |||
AK177985.1 | - | 3 | 166 | |||
BC015259.1 | - | 4 | 222 | 4238768 | ||
BY270130.1 | - | 4 | 241 | |||
U29156.1 | - | 4 | 228 |