Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ereg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007950.2 | NP_031976.1 | 2 | 265 | ||
GenBank mRNA | BB612312.1 | - | 2 | 260 | ||
BC027838.1 | - | 2 | 241 | 5325124 | ||
CJ056327.1 | - | 2 | 266 | |||
D30782.1 | - | 2 | 260 |