Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007952.2 | NP_031978.2 | 4 | 564 | ||
GenBank mRNA | AK088721.1 | - | 4 | 524 | ||
AK132459.1 | - | 4 | 528 | |||
AK133799.1 | - | 4 | 523 | |||
AK167807.1 | - | 4 | 517 | |||
AK169611.1 | - | 4 | 518 | |||
BC003285.1 | - | 4 | 483 | 3582199 | ||
BC033439.1 | - | 4 | 458 | 4016310 | ||
BQ714617.1 | - | 564 | ||||
M73329.1 | - | 4 | 503 |