Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007964.2 | NP_031990.2 | 3 | 469 | ||
GenBank mRNA | AK134294.1 | - | 3 | 670 | ||
AK162905.1 | - | 3 | 729 | |||
AK165332.1 | - | 4 | 462 | |||
AK165885.1 | - | 3 | 422 | |||
BC030056.1 | - | 4 | 476 | |||
BI737814.1 | - | 3 | 393 | |||
U53586.1 | - | 3 | 469 |