Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271616.1 | NP_001258545.1 | 3 | 309 | ||
NM_008009.4 | NP_032035.2 | 2 | 259 | |||
GenBank mRNA | AF065441.1 | - | 1 | 152 | ||
AK146625.1 | - | 2 | 224 | |||
BC065774.1 | - | 2 | 222 | 3975150 | ||
DV059653.1 | - | 3 | 291 | |||
U49641.1 | - | 1 | 1257 |