Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007918.3 | NP_031944.3 | 3 | 824 | ||
GenBank mRNA | AK013033.1 | - | 750 | |||
AK167836.1 | - | 3 | 825 | |||
AK169979.1 | - | 3 | 770 | |||
AK170031.1 | - | 3 | 774 | |||
BC002045.1 | - | 3 | 792 | 3592595 |